Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
Thermo Scientific Chemicals
Iron foil, 0.5mm (0.02in) thick, 99.99% (metals basis)
Form: approx. 2.5g/25x25mm | CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Catalog NumberQuantity
ALF040495.FF25 x 25 mm
Catalog number ALF040495.FF
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 25 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialIron
ColorGray
CAS7439-89-6
Health Hazard 1H500
AppearanceGray foil