Polyethyleneimine, branched, M.W. 10,000, 99%
Polyethyleneimine, branched, M.W. 10,000, 99%
Polyethyleneimine, branched, M.W. 10,000, 99%
Polyethyleneimine, branched, M.W. 10,000, 99%
Polyethyleneimine, branched, M.W. 10,000, 99%
Thermo Scientific Chemicals
Polyethyleneimine, branched, M.W. 10,000, 99%
CAS: 9002-98-6 | ((C2H5N)zC2H4N)y(C2H5N)x | 43.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF040331.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPolyethyleneimine, branched
ColorYellow
Name NoteM.W. 10,000
CAS9002-98-6
Physical FormLiquid
View more
Polyethyleneimine, branched, M.W. 10,000 acts as an auxiliary agent. It is used in paper-making, water treatment, detergents, adhesives and cosmetics. It is investigated as non-viral vector carriers in research.