Neodymium(III) sulfate octahydrate, 99.9% (REO)
Neodymium(III) sulfate octahydrate, 99.9% (REO)
Neodymium(III) sulfate octahydrate, 99.9% (REO)
Neodymium(III) sulfate octahydrate, 99.9% (REO)
Thermo Scientific Chemicals
Neodymium(III) sulfate octahydrate, 99.9% (REO)
CAS: 13477-91-3 | Nd2(SO4)3·8H2O
Have Questions?
Catalog NumberQuantity
ALF040253.1425 g
Catalog number ALF040253.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNeodymium(III) sulfate octahydrate
Melting Point1176°C
Name NoteRare Earth Oxides Content
CAS13477-91-3
ColorPink
View more
It is used as catalyst, glass, crystal and capacitors, in CRT displays to enhance contrast between reds and greens.