Neodymium(III) oxide, REacton™, 99% (REO)
Neodymium(III) oxide, REacton™, 99% (REO)
Neodymium(III) oxide, REacton™, 99% (REO)
Neodymium(III) oxide, REacton™, 99% (REO)
Thermo Scientific Chemicals
Neodymium(III) oxide, REacton™, 99% (REO)
CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF040252.36
Price (MYR)
4,127.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
4,127.00
EA
Add to cart
Specifications
Chemical Name or MaterialNeodymium(III) oxide
Melting Point2272°C
CAS1313-97-9
Recommended StorageAmbient temperatures
Physical FormPowder
View more
ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst.Neodymium(III) oxide is an inorganic compound containing inner transition metal neodymium from lanthanides series in +3 oxidation state and oxide in 2:3 ratio. It adopts a hexagonal crystal structure. It is also known as neodymium oxide, neodymiumtrioxide, neodymium(3+)oxide, neodymium sesquioxide and dineodymiumtrioxide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Ceramic capacitors, coloring glass, carbon arc-light electrodes, color TV tubes, dehydrogenation catalyst
RUO – Research Use Only