Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
Thermo Scientific Chemicals
Ammonium fluoride hydrate, Puratronic™, 99.995% (metals basis)
CAS: 12449-35-3 | FH4N | 37.04 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF040216.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium fluoride hydrate
CAS12449-35-3
ColorWhite
Health Hazard 1H301+H311+H331
Health Hazard 2GHS H Statement
H301-H311-H331
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
View more
Ammonium fluoride is used for etching of glass, preservation of food, in printing textile, and as mothproofing agent. It is used in chemical analysis and in brewing. It is used in the synthesis of NaHoF4 and NaEuF4, which are important in solid state lasers as well as photoluminescence. The fluoride ion is used as a base and nucleophile in organic synthesis. Ammonium fluoride providee safe deprotection strategy for the tert-butyldimethylsilyl (TBS) group during RNA synthesis.