Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
Thermo Scientific Chemicals
Cerium(IV) ammonium sulfate dihydrate, ACS, 94% min
CAS: 10378-47-9 | (NH4)4Ce(SO4)4·2H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 kg
250 g
Catalog number ALF040194.A1
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialCerium(IV) ammonium sulfate dihydrate
Melting Point130°C
Name NoteFor analysis ACS
CAS10378-47-9
ColorYellow
View more
Ammonium cerium(IV) sulfate dihydrate is an oxidizing agent in organic synthesis and it is used as an analytical reagent, in thin-layer chromatography for the detection of anti-depressant drugs in human blood and urine samples. It is used to wet etch the Co catalyst and synthesis of carbon Nanotubes. It is used in medical and pharmaceutical industry.