Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Indium(III) nitrate hydrate, 99.99% (metals basis)
Thermo Scientific Chemicals
Indium(III) nitrate hydrate, 99.99% (metals basis)
CAS: 207398-97-8 | InN3O9 | 300.83 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF040185.18
Price (MYR)
2,000.00
EA
Add to cart
Quantity:
50 g
Request bulk or custom format
Price (MYR)
2,000.00
EA
Add to cart
Specifications
Chemical Name or MaterialIndium(III) nitrate hydrate
Melting Point100°C-H2O
CAS207398-97-8
ColorWhite
Health Hazard 1H272-H315-H319
View more
Indium(III) nitrate hydrate acts as an oxidizing agent. It is used in the manufacture of coordinated polymer films which are used in the preparation of indium(III) oxide nanoparticles. It is used in the synthesis of other indium complexes.