Phosphotungstic acid hydrate
Phosphotungstic acid hydrate
Thermo Scientific Chemicals
Phosphotungstic acid hydrate
CAS: 12067-99-1 | P2O5·24WO3·xH2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF040116.36
Price (MYR)
4,012.00
EA
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (MYR)
4,012.00
EA
Add to cart
Specifications
AppearanceWhite to off-white powder
Chemical Name or MaterialPhosphotungstic acid hydrate
ColorWhite
CAS12067-99-1
Health Hazard 1H314-H335
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Phosphotungstic acid is used as a catalyst for the hydrolysis of propene to prepare 2-propanol. It is also involved in heterogeneous catalysis reaction to prepare propene from 2-propanol and methanol from hydrocarbons. Its ethanolic solution is used in biology as well as to precipitate proteins. It is utilized to precipitate different types of dye such as basic dyes and triphenylmethane dyes. In histology, it is used for staining of cell specimens. It is used in the gravimetric analysis of potassium in analytical chemistry.
Solubility
Soluble in water, alcohol and ether.
Notes
Incompatible with strong bases, strong oxidizing agents.
RUO – Research Use Only
General References:
  1. Zhao, S.; Xu, G.; Chang, J.; Chang, C.; Bai, J.; Fang, S.; Liu, Z. Direct Production of Ethyl Levulinate from Carbohydrates Catalyzed by H-ZSM-5 Supported Phosphotungstic Acid. Bioresources 2015, 10 (2), 2223-2234.
  2. Villa, K.; Murcia-López, S.; Andreu, T.; Morante, J. R. Mesoporous WO3 photocatalyst for the partial oxidation of methane to methanol using electron scavengers. Appl. Catal., B 2015, 163, 150-155.