Nickel Aluminum
Nickel Aluminum
Nickel Aluminum
Thermo Scientific Chemicals
Nickel Aluminum
Wear resistant coatings | CAS: 12635-27-7 | AlNi | 85.675 g/mol
Have Questions?
Catalog NumberQuantity
ALF039828.36500 g
Catalog number ALF039828.36
Price (MYR)
-
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel Aluminum
ColorBlack to Gray
CAS12635-27-7
Health Hazard 1H261-H350
Health Hazard 2GHS H Statement
H228-H261-H372-H351-H317
Flammable solid.
In contact with water releases flammable gas.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
View more
Nickel Aluminum is used as wear resistant coatings. It is also used for the preparation of Raney™ Nickel catalyst.