Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
Thermo Scientific Chemicals
Iron granules, 1-2mm (0.04-0.08in), 99.98% (metals basis)
CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Catalog NumberQuantity
ALF039708.30250 g
Catalog number ALF039708.30
Price (MYR)
371.00
EA
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (MYR)
371.00
EA
Add to cart
Specifications
Chemical Name or MaterialIron
ColorGray
CAS7439-89-6
Health Hazard 1H500
AppearanceGray color granules