Iron foil, 0.127mm (0.005in) thick, 99.5% (metals basis)
Iron foil, 0.127mm (0.005in) thick, 99.5% (metals basis)
Iron foil, 0.127mm (0.005in) thick, 99.5% (metals basis)
Iron foil, 0.127mm (0.005in) thick, 99.5% (metals basis)
Thermo Scientific Chemicals
Iron foil, 0.127mm (0.005in) thick, 99.5% (metals basis)
CAS: 7439-89-6 | Fe | 55.85 g/mol
Have Questions?
Catalog NumberQuantity
ALF039707.FQ25 x 100 mm
Catalog number ALF039707.FQ
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 x 100 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialIron foil
Name Note0.127mm (0.005 in.) thick
CAS7439-89-6
Health Hazard 1H500