Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
Thermo Scientific Chemicals
Nickel gauze, 40 mesh woven from 0.13mm (0.005in) dia wire
CAS: 7440-02-0 | Ni | 58.69 g/mol
Have Questions?
Catalog NumberQuantity
ALF039703.WM600 x 1200 mm
Catalog number ALF039703.WM
Price (MYR)
5,127.00
EA
Add to cart
Quantity:
600 x 1200 mm
Request bulk or custom format
Price (MYR)
5,127.00
EA
Add to cart
Specifications
Chemical Name or MaterialNickel gauze
Name NoteWoven from 0.13mm (0.005 in.) dia. wire
CAS7440-02-0
Health Hazard 1H317-H351-H372
Health Hazard 2GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.