Copper powder, -100 mesh, 99.999% (metals basis)
Copper powder, -100 mesh, 99.999% (metals basis)
Copper powder, -100 mesh, 99.999% (metals basis)
Copper powder, -100 mesh, 99.999% (metals basis)
Thermo Scientific Chemicals
Copper powder, -100 mesh, 99.999% (metals basis)
CAS: 7440-50-8 | Cu | 63.55 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF039682.22
Price (MYR)
6,605.00
EA
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (MYR)
6,605.00
EA
Add to cart
Specifications
Chemical Name or MaterialCopper powder
ColorRed Brown
CAS7440-50-8
Health Hazard 1H228-H315-H319-H335-H500
Health Hazard 2GHS H Statement
H228-H319-H335
Flammable solid.