Tungstosilicic acid hydrate, Reagent Grade
Tungstosilicic acid hydrate, Reagent Grade
Tungstosilicic acid hydrate, Reagent Grade
Tungstosilicic acid hydrate, Reagent Grade
Thermo Scientific Chemicals
Tungstosilicic acid hydrate, Reagent Grade
CAS: 12027-43-9 | H4O40SiW12 | 2878.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
10 g
Catalog number ALF039651.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialTungstosilicic acid hydrate
Name NoteReagent Grade
CAS12027-43-9
Health Hazard 1H314-H335
Health Hazard 3P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
View more
Catalyst for organic synthesis, minerals separation, reagent for alkaloids.Tungstosilicic acid hydrate is used to prepare ethylacetate and acetic acid from ethylene. It is used in the preparation of silicotungstate butylmethylimidazolium bromide and charge transfer compounds with Keggin anions. It is also used in catalyst preparation, which is useful for catalytic liquid-phase oxidation of xylene. Further, it acts as a mediator in production of hydrogen through electrolysis of water.