Potassium antimonate trihydrate, 94+%
Potassium antimonate trihydrate, 94+%
Potassium antimonate trihydrate, 94+%
Potassium antimonate trihydrate, 94+%
Thermo Scientific Chemicals
Potassium antimonate trihydrate, 94+%
CAS: 12208-13-8 | KSbO3·3H2O
Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
50 g
Catalog number ALF039524.30
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium antimonate trihydrate
CAS12208-13-8
ColorWhite
Health Hazard 1H302+H332
Health Hazard 3P261-P264b-P270-P271-P301+P312-P304+P340-P312-P330-P501c
View more
Potassium antimonyl trihydrate is used as a detecting agent for sodium. It is used as a mordant/ fixing agent in leather and textile dying industries. It is also used as a analytical reagent and flux additive for electroplating.