Potassium orthovanadate, 99.9% (metals basis)
Potassium orthovanadate, 99.9% (metals basis)
Potassium orthovanadate, 99.9% (metals basis)
Potassium orthovanadate, 99.9% (metals basis)
Thermo Scientific Chemicals
Potassium orthovanadate, 99.9% (metals basis)
CAS: 14293-78-8 | K3O4V | 232.232 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF039502.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPotassium orthovanadate
CAS14293-78-8
Health Hazard 1H315-H319-H332-H335
Health Hazard 2GHS H Statement
H331-H315-H319-H335
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
It is employed as in chemical fertilizer industries and used as corrosion inhibitor. It is also used for ammonia synthesis. Used as a catalyst in oil & gas industry.