Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
Nickel(II) phthalocyanine
Thermo Scientific Chemicals
Nickel(II) phthalocyanine
CAS: 14055-02-8 | C32H16N8Ni | 571.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Catalog number ALF039453.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNickel(II) phthalocyanine
CAS14055-02-8
Health Hazard 1H317-H334-H351
Health Hazard 2GHS H Statement
H350-H302-H312-H332
May cause cancer.
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 3P201-P202-P261-P272-P280-P285-P302+P352-P304+P341-P308+P313-P333+P313-P342+P311-P363-P501c