Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
Thermo Scientific Chemicals
Cyclopentadienylmolybdenum tricarbonyl dimer, 98%
CAS: 12091-64-4 | C16H2Mo2O6 | 482.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number ALF039245.06
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 g
Request bulk or custom format
Specifications
Chemical Name or MaterialCyclopentadienylmolybdenum tricarbonyl dimer
CAS12091-64-4
Health Hazard 1H302+H312+H332
Health Hazard 2GHS H Statement
H300-H330
Fatal if swallowed.
Fatal if inhaled.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P312-P330-P363-P501c
View more
It is widely used as a chemical intermediate in the laboratories, manufacture purposes, chemical and pharmaceutical intemediate.