Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Bis(1,5-cyclooctadiene)nickel(0)
Thermo Scientific Chemicals
Bis(1,5-cyclooctadiene)nickel(0)
CAS: 1295-35-8 | C16H24Ni | 275.06 g/mol
Have Questions?
Catalog NumberQuantity
ALF039234.042 g
Catalog number ALF039234.04
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 g
Request bulk or custom format
Specifications
Chemical Name or MaterialBis(1,5-cyclooctadiene)nickel(0)
CAS1295-35-8
Health Hazard 1H228-H317-H350
Health Hazard 2GHS H Statement
H228-H350-H317
Flammable solid.
May cause cancer.
May cause an allergic skin reaction.
Health Hazard 3P201-P202-P210-P240-P241-P261-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P370+P378q-P501c
View more
Bis(1,5-cyclooctadiene)nickel(0) is used as a catalyst for the cycloaddition of 1,3-dienes and is used to catalyze the addition of allyl phenyl sulfide to alkynes leading to 1,4-dienes. Also, it is involved as a catalyst for asymmetric alpha-arylation and heteroarylation of ketones with chloroarenes, stereoselective borylative ketone-diene coupling, cycloaddition of benzamides with internal alkynes and methyl carboxylation of homopropargylic alcohols.