1-Chlorobutane, HPLC Grade, 99.5+%
1-Chlorobutane, HPLC Grade, 99.5+%
1-Chlorobutane, HPLC Grade, 99.5+%
1-Chlorobutane, HPLC Grade, 99.5+%
Thermo Scientific Chemicals
1-Chlorobutane, HPLC Grade, 99.5+%
Maximum level of impurities: Evaporation residue 1ppm, H2O 0.02% | CAS: 109-69-3 | C4H9Cl | 92.57 g/mol
Have Questions?
Catalog NumberQuantity
ALF039198.AK250 mL
Catalog number ALF039198.AK
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
250 mL
Request bulk or custom format
Specifications
Chemical Name or Material1-Chlorobutane
Name NoteFiltered through 0.2μ filters.
CAS109-69-3
Health Hazard 1H225-H304
Health Hazard 3P210-P233-P240-P241-P242-P243-P280-P301+P310-P303+P361+P353-P331-P370+P378q-P501c
View more
1-Chlorobutane is used as intermediates and solvents. It is used for HPLC, spectrophotometry and environmental testing. It is used as alkylating reagent in organic and organometallic compound synthesis. It is also employed as an antihelmintic in veterinary medicine.