Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol
Thermo Scientific Chemicals
Tungsten(VI) ethoxide, 99.8% (metals basis), 5% w/v in ethanol
CAS: 62571-53-3 | C12H30O6W | 454.21 g/mol
Have Questions?
Catalog NumberQuantity
ALF039170.AC25 mL
Catalog number ALF039170.AC
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialTungsten(VI) ethoxide
CAS62571-53-3
Health Hazard 1H225-H319-H335
Health Hazard 2GHS H Statement
H225
Highly flammable liquid and vapor.
Health Hazard 3P210-P233-P240-P241-P242-P243-P264b-P280-P303+P361+P353-P305+P351+P338-P370+P378q-P501c
View more
Tungsten(VI) ethoxide is used in nano-materials, thin films, display and imaging, optics and glasses. It is also used for x-ray screen phosphors, fireproofing fabrics and in gas sensors. It acts as an intermediate for sol-gel coatings.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For Manufacturing and Laboratory Use. Not for beverage use