1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
Thermo Scientific Chemicals
1,4-Dioxane, ACS, 99+%, stab. with 1-3ppm BHT
CAS: 123-91-1 | C4H8O2 | 88.106 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF039118.K2
Price (MYR)
696.00
EA
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (MYR)
696.00
EA
Add to cart
Specifications
Chemical Name or Material1, 4-Dioxane
Melting Point11.8°C
Name NoteFor analysis ACS
CAS123-91-1
ColorColorless
View more
1,4-Dioxane is used as a stabilizer for 1,1,1-trichloroethane and chlorinated solvents. It acts as a solvent for cellulose acetate, ethyl cellulose, benzyl cellulose, resins, oils, waxes, dyes, other organic and inorganic compounds. It acts as a substitute for tetrahydrofuran due to its higher boiling point and lower toxicity. Furthermore, it is used as an internal standard for proton NMR spectroscopy in deuterium oxide. It plays an important role in liquid scintillation counting in biological procedures and as a wetting and dispersing agent in textile processing. It is involved in the preparation of dimethylmagnesium by reacting with methylmagnesium bromide and in the preparation of histological sections for microscopic examination.