Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
Thermo Scientific Chemicals
Silicon plate, 6.35mm (0.25in) thick, 99.999% (metals basis)
CAS: 7440-21-3 | Si | 28.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 x 100 mm
25 x 25 mm
50 x 50 mm
Catalog number ALF039112.GH
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 x 100 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialSilicon plate
Name Note6.35mm (0.25 in.) thick
CAS7440-21-3
Recommended StorageAmbient temperatures
EINECS Number231-130-8