Iodine monochloride
Iodine monochloride
Iodine monochloride
Thermo Scientific Chemicals
Iodine monochloride
CAS: 7790-99-0 | ClI | 162.35 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number ALF039104.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Boiling Point96°C to 98°C
Chemical Name or MaterialIodine monochloride
Melting Point25°C to 27°C
CAS7790-99-0
Health Hazard 1H300-H311-H314-H335-H373
View more
ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.