Glycerol, ultrapure, HPLC Grade
Glycerol, ultrapure, HPLC Grade
Glycerol, ultrapure, HPLC Grade
Glycerol, ultrapure, HPLC Grade
Thermo Scientific Chemicals
Glycerol, ultrapure, HPLC Grade
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Catalog NumberQuantity
ALF038988.K21 L
Catalog number ALF038988.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol
Name NoteHPLC Grade
CAS56-81-5
Health Hazard 1H319: Causes serious eye irritation.
Melting Point18°C
View more
Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production.