Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
Thermo Scientific Chemicals
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable
CAS: 1336-21-6 | H5NO | 35.05 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF038714.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Chemical Identifiers
CAS1336-21-6
Specifications
Chemical Name or MaterialBuffer solution
CAS1336-21-6
ColorColorless
Recommended StorageAmbient temperatures
TSCAYes
View more
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.