Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
Thermo Scientific Chemicals
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.
CAS: 34722-90-2 | C27H27Br2NaO5S | 646.37 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
100 mL
Catalog number ALF038701.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialBromothymol Blue sodium salt
CAS34722-90-2
Recommended StorageAmbient temperatures
TSCAYes
Molecular FormulaC27H27Br2NaO5S