View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 x 200 mm
Request bulk or custom format
Specifications
Chemical Name or MaterialAluminum foil
Name Note0.038 ±0.007mm (0.0015 ±0.0003 in.) thick
CAS7429-90-5
Recommended StorageAmbient temperatures
EINECS Number231-072-3