Semiquantitative Standard 2, Specpure™
Semiquantitative Standard 2, Specpure™
Thermo Scientific Chemicals
Semiquantitative Standard 2, Specpure™
Have Questions?
Catalog NumberQuantity
ALF036770.AE100 mL
Catalog number ALF036770.AE
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialSemiquantitative Standard 2
Health Hazard 1Specific target organ toxicity after single exposure (category 3), Corrosive to metals (category 1), Skin corrosion/irritation (category 1)
Health Hazard 2May cause respiratory irritation, May be corrosive to metals, Causes severe skin burns and eye damage
Recommended StorageAmbient temperatures
DOT InformationTransport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, OXIDIZING, N.O.S.
View more
Semiquantitative Standard 2 is utilized as standard solution in analytical chemistry.