3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
3-Methyl-1-butanol, ACS, 98.5+%
Thermo Scientific Chemicals
3-Methyl-1-butanol, ACS, 98.5+%
CAS: 123-51-3 | C5H12O | 88.15 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF036716.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or Material3-Methyl-1-butanol
CAS123-51-3
Health Hazard 1H226-H315-H319-H332-H335
Health Hazard 2GHS H Statement
H226-H332-H335
Flammable liquid and vapor.
Harmful if inhaled.
May cause respiratory irritation.
Health Hazard 3P210-P233-P235-P240-P241-P242-P243-P260-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P314-P332+P313-P363-P370+P378q-P501c
View more
3-Methyl-1-butanol is used as an active ingredient in banana oil production and Kovac's reagent. It acts as an antifoaming agent. It serves as an intermediate for the manufacture of corrosion inhibitors, flotation aids, fuel and lubricating oil additives, polyvinyl chloride stabilizers, pharmaceuticals, cosmetics, plasticizers, rubber auxiliaries and paint solvents.