4-Dimethylaminobenzaldehyde, ACS
4-Dimethylaminobenzaldehyde, ACS
4-Dimethylaminobenzaldehyde, ACS
4-Dimethylaminobenzaldehyde, ACS
Thermo Scientific Chemicals
4-Dimethylaminobenzaldehyde, ACS
CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF036684.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or Material4-Dimethylaminobenzaldehyde
CAS100-10-7
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P264b-P270-P301+P312-P330-P501c
View more
4-Dimethylaminobenzaldehyde is used to prepare colorful Schiff base adducts with amines, pyrroles and indoles. It is often used in the Ehrlich reaction, test for the presence of indoles in a sample containing alkaloids and determination of hydrazine.