Citric acid, anhydrous, ACS, 99.5+%
Citric acid, anhydrous, ACS, 99.5+%
Citric acid, anhydrous, ACS, 99.5+%
Citric acid, anhydrous, ACS, 99.5+%
Thermo Scientific Chemicals
Citric acid, anhydrous, ACS, 99.5+%
CAS: 77-92-9 | C6H8O7 | 192.12 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
100 g
500 g
Catalog number ALF036664.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialCitric Acid
Name NoteFor analysis ACS
CAS77-92-9
Melting Point153°C to 154°C
Health Hazard 1H319-H335-H500
View more
Citric acid is the most widely used organic acid and pH-control agent in foods, beverages, pharmaceuticals and technical applications. Used in pharmaceutical preparations, especially in effervescent tablets. It is used as a natural preservative.