Glycerol, ACS, 99.5+%
Glycerol, ACS, 99.5+%
Glycerol, ACS, 99.5+%
Glycerol, ACS, 99.5+%
Thermo Scientific Chemicals
Glycerol, ACS, 99.5+%
CAS: 56-81-5 | C3H8O3 | 92.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
Catalog number ALF036646.K2
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
1 L
Request bulk or custom format
Specifications
Chemical Name or MaterialGlycerol
CAS56-81-5
Health Hazard 1H319: Causes serious eye irritation.
Melting Point18°C
Recommended StorageAmbient temperatures
View more
Glycerol is widely used in cosmetics, especially for production of soaps and lotions, solvents and emulsifiers. It may also be found in inks, adhesives and pharmaceuticals. It plays an important role in the manufacture of plastics and resins. Glycerol bonded with silica gel plays an important role for effective removal of traces of Co(II) from aqueous solutions. Also useful for optical clearing of gastric tissue.