Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Formic acid, ACS, 96+%
Thermo Scientific Chemicals
Formic acid, ACS, 96+%
CAS: 64-18-6 | CH2O2 | 46.025 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
2 kg
500 g
Catalog number ALF036617.A3
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
2 kg
Request bulk or custom format
Specifications
Chemical Name or MaterialFormic acid
Name NoteFor analysis ACS
CAS64-18-6
Melting Point7°C to 9°C
Boiling Point100°C to 101°C
View more
Used as a hydrogen source, as a reducing agent and is used in tanning and dye fixing.