Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
Thermo Scientific Chemicals
Manganese(II) chloride tetrahydrate, ACS, 98.0-101.0%
CAS: 13446-34-9 | Cl2H8MnO4 | 197.90 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036526.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese(II) chloride tetrahydrate
Melting Point58°C
CAS13446-34-9
ColorPink
Health Hazard 1H302-H318-H373
View more
It is used in the production of dry cell batteries. It is a precursor material for antiknock compound methylcyclopentadienyl manganese tricarbony. It is also useful for coupling reactions as well as organic synthesis. It is a catalyst for fast and slow dehydrogenation of ball milled lithium alanate.