N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
N,N-Dimethyl-1-naphthylamine, 99%
Thermo Scientific Chemicals
N,N-Dimethyl-1-naphthylamine, 99%
CAS: 86-56-6 | C12H13N | 171.243 g/mol
Have Questions?
Catalog NumberQuantity
ALF036438.AA5 mL
Catalog number ALF036438.AA
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
5 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialN,N-Dimethyl-1-naphthylamine
CAS86-56-6
Health Hazard 1H302-H315-H319-H335
Health Hazard 2GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
View more
N,N-Dimethyl-1-naphthylamine is used in the nitrate reductase test to form a red precipitate of Prontosil by reacting with a nitrite-sulfanilic acid complex. And is also used to detection and determination of nitrites.