Hydroxylamine hydrochloride, ACS, 96+%
Hydroxylamine hydrochloride, ACS, 96+%
Hydroxylamine hydrochloride, ACS, 96+%
Hydroxylamine hydrochloride, ACS, 96+%
Thermo Scientific Chemicals
Hydroxylamine hydrochloride, ACS, 96+%
A Monoamine Oxidase Inhibitor; Inhibits Platelet Aggregation | CAS: 5470-11-1 | ClH4NO | 69.49 g/mol
Have Questions?
Catalog NumberQuantity
ALF036416.22100 g
Catalog number ALF036416.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialHydroxylamine hydrochloride
Melting Point152°C (decomposition)
Name NoteFor analysis ACS
CAS5470-11-1