L-(+)-Tartaric acid, ACS
L-(+)-Tartaric acid, ACS
L-(+)-Tartaric acid, ACS
L-(+)-Tartaric acid, ACS
Thermo Scientific Chemicals
L-(+)-Tartaric acid, ACS
Useful as a resolving agent | CAS: 87-69-4 | C4H6O6 | 150.09 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036405.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialL-(+)-Tartaric acid
pH2.1
CAS87-69-4
Melting Point168°C to 170°C
Health Hazard 1H318-H500
View more
L-(+)-Tartaric acid is widely utilized in pharmaceutical industries. It is used in soft drinks, confectionaries, food products, gelatin desserts and buffering agents. It forms a compound, TiCl2(O-i-Pr)2 with Diels-Alder catalyst and act as a chelate agent in metal industries. Owing to its efficient chelating property towards metal ions, it is used in the farming and metal industries for complexing micronutrients and for cleaning metal surfaces, respectively.