Germanium(IV) oxide, 99.99% (metals basis)
Germanium(IV) oxide, 99.99% (metals basis)
Germanium(IV) oxide, 99.99% (metals basis)
Germanium(IV) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals
Germanium(IV) oxide, 99.99% (metals basis)
Used in Phosphors, Transistors and Diodes, and Infrared Transmitting Glass | CAS: 1310-53-8 | GeO2 | 104.628 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
50 g
2 g
10 g
Catalog number ALF036382.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Chemical Name or MaterialGermanium(IV) oxide
Melting Point1086°C
CAS1310-53-8
ColorWhite
Health Hazard 1H302+H332
View more
Used in phosphors, transistors and diodes, and infrared transmitting glass.