D-(-)-Isoascorbic acid, 98%
D-(-)-Isoascorbic acid, 98%
D-(-)-Isoascorbic acid, 98%
D-(-)-Isoascorbic acid, 98%
Thermo Scientific Chemicals
D-(-)-Isoascorbic acid, 98%
CAS: 89-65-6 | C6H7NaO6 | 198.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF036366.22100 g
Catalog number ALF036366.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialD-(-)-Isoascorbic acid
CAS89-65-6
Melting Point169°C to 172°C (decomposition)
Recommended StorageAmbient temperatures
RTECS NumberKF3015000
View more
D-(-)-Isoascorbic acid is used as antioxidant especially in brewing industry, reducing agent in photography. And it is also used in food industry, as food additives.