Silicon(IV) oxide, 99.99% (metals basis)
Silicon(IV) oxide, 99.99% (metals basis)
Silicon(IV) oxide, 99.99% (metals basis)
Silicon(IV) oxide, 99.99% (metals basis)
Thermo Scientific Chemicals
Silicon(IV) oxide, 99.99% (metals basis)
CAS: 7631-86-9 | O2Si | 60.08 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF036346.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Boiling Point2230°C
Chemical Name or MaterialSilicon(IV) oxide
Melting Point1710°C
CAS7631-86-9
Recommended StorageAmbient temperatures
View more
Silica is used primarily in the production of glass for windows, drinking glasses, beverage bottles. The majority of optical fibers for telecommunication are also made from silica. It is a primary raw material for many ceramics such as earthenware, stoneware, and porcelain. The main ingredient in sand casting for the manufacture of a large number of metallic components in engineering and other applications. Silica is a common additive in the production of foods. An estimated 95% of silicon dioxide produced is consumed in the construction industry.