View Price:Sign InSign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese
CAS7439-96-5
Health Hazard 1H228-H319
Health Hazard 2Take precautionary measures against static discharges.
Health Hazard 3P210-P240-P241-P264b-P280-P305+P351+P338-P370+P378q