Antimony(III) chloride, ACS, 99.0% min
Antimony(III) chloride, ACS, 99.0% min
Antimony(III) chloride, ACS, 99.0% min
Antimony(III) chloride, ACS, 99.0% min
Thermo Scientific Chemicals
Antimony(III) chloride, ACS, 99.0% min
CAS: 10025-91-9 | Cl3Sb | 228.11 g/mol
Have Questions?
Catalog NumberQuantity
ALF036281.1850 g
Catalog number ALF036281.18
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
50 g
Request bulk or custom format
Specifications
Boiling Point223.5°C
Chemical Name or MaterialAntimony(III) chloride
Melting Point73.4°C
Name NoteFor analysis ACS
CAS10025-91-9
View more
Antimony(III) chloride is engaged to detect certain compounds, such as cholesterols and carotenoids. It is likewise employed as a dye mordant and a fabric flame retardant. It is used as inorganic and organic chlorination catalyst. It is used to coat the steel to make bronze, used as a cutting material of antimony salt and as a preservative in medicine.