Thulium(III) oxide, 99.9% (REO)
Thulium(III) oxide, 99.9% (REO)
Thulium(III) oxide, 99.9% (REO)
Thulium(III) oxide, 99.9% (REO)
Thermo Scientific Chemicals
Thulium(III) oxide, 99.9% (REO)
CAS: 12036-44-1 | O3Tm2 | 385.87 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
10 g
2 g
Catalog number ALF036279.09
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
10 g
Request bulk or custom format
Specifications
Chemical Name or MaterialThulium(III) oxide
Melting Point2425°C
CAS12036-44-1
ColorGreen
Health Hazard 1Warning