Neodymium(III) oxide, 99.9% (REO)
Neodymium(III) oxide, 99.9% (REO)
Neodymium(III) oxide, 99.9% (REO)
Neodymium(III) oxide, 99.9% (REO)
Thermo Scientific Chemicals
Neodymium(III) oxide, 99.9% (REO)
CAS: 1313-97-9 | Nd2O3 | 336.48 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number ALF036275.22
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
100 g
Request bulk or custom format
Specifications
Chemical Name or MaterialNeodymium(III) oxide
Melting Point2272°C
CAS1313-97-9
ColorBlue
Recommended StorageAmbient temperatures
View more
Neodymium is used in coloring glasses and for doping some glass lasers since it transmits 90% of the blue, green, and red light rays. Neodymium compounds are used in glass coloring, permanent magnets, capacitors, infrared filters, resonators, Catalysis (diene polymerization), laser materials. Neodymium(III) oxide is used to dope glass, including sunglasses, to make solid-state lasers, and to color glasses and enamels.