Ethanolamine, ACS, 99+%
Ethanolamine, ACS, 99+%
Ethanolamine, ACS, 99+%
Ethanolamine, ACS, 99+%
Thermo Scientific Chemicals
Ethanolamine, ACS, 99+%
CAS: 141-43-5 | C2H7NO | 61.08 g/mol
Have Questions?
Catalog NumberQuantity
ALF036260.AP500 mL
Catalog number ALF036260.AP
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 mL
Request bulk or custom format
Specifications
Chemical Name or MaterialEthanolamine
Name NoteFor analysis ACS
CAS141-43-5
Health Hazard 1H227-H302+H312+H332-H314-H335
Health Hazard 2GHS H Statement
H314-H302-H312-H332-H335-H227
View more
It is also used in messenger molecules such as palmitoylethanolamide, which have an effect on CB1 receptors. Ethanolamine is used as a scrubber for the removal of methyl bromide. Polyaniline doping ethanolamine is used for modified electrode preparation to measure uric acid in human body with the presence of antiscorbutic acid.