Resorcinol, ACS, 99.0-100.5%
Resorcinol, ACS, 99.0-100.5%
Resorcinol, ACS, 99.0-100.5%
Resorcinol, ACS, 99.0-100.5%
Thermo Scientific Chemicals
Resorcinol, ACS, 99.0-100.5%
CAS: 108-46-3 | C6H6O2 | 110.112 g/mol
Have Questions?
Catalog NumberQuantity
ALF036248.1425 g
Catalog number ALF036248.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialResorcinol
Name NoteFor analysis ACS
CAS108-46-3
Health Hazard 1H301-H315-H319-H500
Health Hazard 2GHS H Statement
H302-H315-H319
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
View more
Resorcinol is used in the synthesis of pharmaceuticals and various organic compounds. It is likewise used in plasticizers and diazo dyes. It can be applied in the qualitative determination of ketoses in the Seliwanoff test. Resorcinol-formaldehyde resin has shown useful application in endodontic therapy in russian red. It is an indispensable element for spectrophotometric determination of nitrate concentrations in seawater.