Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Ammonium carbonate, ACS, NH{3} 30.0+%
Thermo Scientific Chemicals
Ammonium carbonate, ACS, NH{3} 30.0+%
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue | CAS: 506-87-6 | CH8N2O3 | 96.086 g/mol
Have Questions?
Catalog NumberQuantity
ALF036229.36500 g
Catalog number ALF036229.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium carbonate
Melting Point60° (sublimation)
CAS506-87-6
Health Hazard 1H302
Health Hazard 2GHS H Statement
H302-H315-H319-H335
View more
In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compoundsAmmonium carbonate is widely utilized as a leavening agent in lebkuchen, cookies and flat biscuits. It finds an important application as an emetic and an active ingredient
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • In baking powder, smelling salts, as an analytical lab reagent, manufacturing of rubber materials, casein glue, and fire extinguishing compounds
RUO – Research Use Only