Ammonium oxalate monohydrate, ACS, 99.0-101.0%
Ammonium oxalate monohydrate, ACS, 99.0-101.0%
Ammonium oxalate monohydrate, ACS, 99.0-101.0%
Ammonium oxalate monohydrate, ACS, 99.0-101.0%
Thermo Scientific Chemicals
Ammonium oxalate monohydrate, ACS, 99.0-101.0%
In detinning of iron, metal polishing | CAS: 6009-70-7 | C2H10N2O5 | 142.111 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
Catalog number ALF036228.36
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
500 g
Request bulk or custom format
Specifications
Chemical Name or MaterialAmmonium oxalate
Melting Point70°C (decomposition)
Name NoteMonohydrate
CAS6009-70-7
Health Hazard 1H302+H312-H320-H335-H372
View more
In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metalsAmmonium oxalate monohydrate is widely utilized as a buffering reagent and a dispersant to determine the surface interaction of platelets. It finds an application to study
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • In detinning of iron, metal polishing, determination of lead, calcium, and rare earth metals
RUO – Research Use Only