Manganese pieces, irregular, 99.95% (metals basis)
Manganese pieces, irregular, 99.95% (metals basis)
Manganese pieces, irregular, 99.95% (metals basis)
Manganese pieces, irregular, 99.95% (metals basis)
Thermo Scientific Chemicals
Manganese pieces, irregular, 99.95% (metals basis)
For Vacuum Deposition | CAS: 7439-96-5 | Mn | 54.94 g/mol
Have Questions?
Catalog NumberQuantity
ALF036221.1425 g
Catalog number ALF036221.14
View Price:Sign in to see your account pricing. Need an account? Register with us today.
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialManganese
ColorGray
CAS7439-96-5
AppearanceGray irregularly shaped pieces
Recommended StorageAmbient temperatures
View more
For vacuum deposition